Anti-SFTPB Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP41411_P050
AVIVARP41411_P050EA 401 EUR
AVIVARP41411_P050
Anti-SFTPB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SFTPB (surfactant protein B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Dog, Guinea pig, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR