Anti-UGT1A6 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP41458_P050
AVIVARP41458_P050EA 401 EUR
AVIVARP41458_P050
Anti-UGT1A6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UGT1A6 (UDP glucuronosyltransferase 1 family, polypeptide A6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen