Anti-ACP5 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP41471_P050
AVIVARP41471_P050EA 401 EUR
AVIVARP41471_P050
Anti-ACP5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACP5 (acid phosphatase 5, tartrate resistant)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen