Anti-BHMT Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP41475_T100
AVIVARP41475_T100EA 349 EUR
AVIVARP41475_T100
Anti-BHMT Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BHMT (betaine--homocysteine S-methyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR