Anti-MARCO Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP41580_P050
AVIVARP41580_P050EA 401 EUR
AVIVARP41580_P050
Anti-MARCO Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MARCO (macrophage receptor with collagenous structure)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human