Anti-STS Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP41709_P050
AVIVARP41709_P050EA 401 EUR
AVIVARP41709_P050
Anti-STS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STS (steroid sulfatase (microsomal), isozyme S)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen