Anti-SIGLEC6 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP41925_P050
AVIVARP41925_P050EA 401 EUR
AVIVARP41925_P050
Anti-SIGLEC6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SIGLEC6 (sialic acid binding Ig-like lectin 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human