Anti-APCS Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP41962_T100
AVIVARP41962_T100EA 349 EUR
AVIVARP41962_T100
Anti-APCS Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: APCS (amyloid P component, serum)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR