Anti-EPS8L1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP42492_P050
AVIVARP42492_P050EA 401 EUR
AVIVARP42492_P050
Anti-EPS8L1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EPS8L1 (EPS8-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen