Anti-QRSL1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP42512_P050
AVIVARP42512_P050EA 401 EUR
AVIVARP42512_P050
Anti-QRSL1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: QRSL1 (glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen