Anti-PRR5L Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP42638_P050
AVIVARP42638_P050EA 401 EUR
AVIVARP42638_P050
Anti-PRR5L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRR5L (proline rich 5 like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen