Anti-S1PR5 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP42661_T100
AVIVARP42661_T100EA 349 EUR
AVIVARP42661_T100
Anti-S1PR5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: S1PR5 (sphingosine-1-phosphate receptor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Dog, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR