Anti-RAG1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP42949_P050
AVIVARP42949_P050EA 401 EUR
AVIVARP42949_P050
Anti-RAG1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAG1 (recombination activating gene 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR