Anti-MARCH2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP42966_P050
AVIVARP42966_P050EA 401 EUR
AVIVARP42966_P050
Anti-MARCH2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MARCH2 (membrane-associated ring finger (C3HC4) 2, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR