Anti-UBE2K Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP43049_P050
AVIVARP43049_P050EA 401 EUR
AVIVARP43049_P050
Anti-UBE2K Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UBE2K (ubiquitin-conjugating enzyme E2K)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen