Anti-MARCH2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP43219_P050
AVIVARP43219_P050EA 401 EUR
AVIVARP43219_P050
Anti-MARCH2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MARCH2 (membrane-associated ring finger (C3HC4) 2, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen