Anti-SLC30A3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP43848_P050
AVIVARP43848_P050EA 401 EUR
AVIVARP43848_P050
Anti-SLC30A3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC30A3 (solute carrier family 30 (zinc transporter), member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen