Anti-SLC7A7 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP43863_P050
AVIVARP43863_P050EA 401 EUR
AVIVARP43863_P050
Anti-SLC7A7 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC7A7 (solute carrier family 7 (amino acid transporter light chain, y+L system), member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen