Anti-SLC7A8 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP43930_T100
AVIVARP43930_T100EA 349 EUR
AVIVARP43930_T100
Anti-SLC7A8 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC7A8 (solute carrier family 7 (amino acid transporter light chain, L system), member 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen