Anti-SLC15A2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP44016_P050
AVIVARP44016_P050EA 401 EUR
AVIVARP44016_P050
Anti-SLC15A2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC15A2 (solute carrier family 15 (oligopeptide transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen