Anti-GPD2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP44281_P050
AVIVARP44281_P050EA 401 EUR
AVIVARP44281_P050
Anti-GPD2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GPD2 (glycerol-3-phosphate dehydrogenase 2 (mitochondrial))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen