Anti-PPIB Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP44364_P050
AVIVARP44364_P050EA 401 EUR
AVIVARP44364_P050
Anti-PPIB Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PPIB (peptidylprolyl isomerase B (cyclophilin B))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen