Anti-FMO3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP44435_P050
AVIVARP44435_P050EA 401 EUR
AVIVARP44435_P050
Anti-FMO3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FMO3 (flavin containing monooxygenase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen