Anti-CLCC1 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP44970_T100
AVIVARP44970_T100EA 349 EUR
AVIVARP44970_T100
Anti-CLCC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CLCC1 (chloride channel CLIC-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR