Anti-KLRC3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP45260_P050
AVIVARP45260_P050EA 401 EUR
AVIVARP45260_P050
Anti-KLRC3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KLRC3 (killer cell lectin-like receptor subfamily C, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human