Anti-LSAMP Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP45277_P050
AVIVARP45277_P050EA 401 EUR
AVIVARP45277_P050
Anti-LSAMP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LSAMP (limbic system-associated membrane protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR