Anti-OMG Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP45327_P050
AVIVARP45327_P050EA 401 EUR
AVIVARP45327_P050
Anti-OMG Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OMG (oligodendrocyte myelin glycoprotein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen