Anti-FURIN Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP45328_P050
AVIVARP45328_P050EA 401 EUR
AVIVARP45328_P050
Anti-FURIN Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FURIN (furin (paired basic amino acid cleaving enzyme))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen