Anti-PI3 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP45340_P050
AVIVARP45340_P050EA 401 EUR
AVIVARP45340_P050
Anti-PI3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PI3 (peptidase inhibitor 3, skin-derived)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

TECHNISCHE DATEN

Antigen-Symbol PI3
Environmentally Preferable
Antigen-Name Peptidase Inhibitor 3
Antigen-Synonyme WAP3|WFDC14|SKALP|cementoin|ESI
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität Human
Wirt Rabbit
Gen-ID 5266
Isotyp
Western blot Yes
Epitop N-terminal
Kreuzadsorption none
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR