Anti-PTPRR Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP45387_P050
AVIVARP45387_P050EA 401 EUR
AVIVARP45387_P050
Anti-PTPRR Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTPRR (protein tyrosine phosphatase, receptor type, R)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR