Anti-SMPD2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP45417_T100
AVIVARP45417_T100EA 349 EUR
AVIVARP45417_T100
Anti-SMPD2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SMPD2 (sphingomyelin phosphodiesterase 2, neutral membrane (neutral sphingomyelinase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR