Anti-SUMO3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP45841_P050
AVIVARP45841_P050EA 401 EUR
AVIVARP45841_P050
Anti-SUMO3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SUMO3 (small ubiquitin-like modifier 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen