Anti-GUCY1B3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP46012_P050
AVIVARP46012_P050EA 401 EUR
AVIVARP46012_P050
Anti-GUCY1B3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GUCY1B3 (guanylate cyclase 1, soluble, beta 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen