Anti-PRIM1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP46014_P050
AVIVARP46014_P050EA 401 EUR
AVIVARP46014_P050
Anti-PRIM1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRIM1 (primase, DNA, polypeptide 1 (49kDa))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen