Anti-ACTR2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP46022_T100
AVIVARP46022_T100EA 349 EUR
AVIVARP46022_T100
Anti-ACTR2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ACTR2 (ARP2 actin-related protein 2 homolog (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen