Anti-NINL Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP46344_P050
AVIVARP46344_P050EA 401 EUR
AVIVARP46344_P050
Anti-NINL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NINL (ninein-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen