Anti-TMED3 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP46845_T100
AVIVARP46845_T100EA 349 EUR
AVIVARP46845_T100
Anti-TMED3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TMED3 (transmembrane emp24 protein transport domain containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR