Anti-FABP3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP47483_P050
AVIVARP47483_P050EA 401 EUR
AVIVARP47483_P050
Anti-FABP3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FABP3 (fatty acid binding protein 3, muscle and heart (mammary-derived growth inhibitor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen