Anti-ZNF397 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP47578_P050
AVIVARP47578_P050EA 401 EUR
AVIVARP47578_P050
Anti-ZNF397 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF397 (zinc finger protein 397)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen