Anti-PA2G4 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP47602_T100
AVIVARP47602_T100EA 349 EUR
AVIVARP47602_T100
Anti-PA2G4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PA2G4 (proliferation-associated 2G4, 38kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR