Anti-TAMM41 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP47716_P050
AVIVARP47716_P050EA 401 EUR
AVIVARP47716_P050
Anti-TAMM41 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TAMM41 (TAM41, mitochondrial translocator assembly and maintenance protein, homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR