Anti-MAFA Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP47762_P050
AVIVARP47762_P050EA 401 EUR
AVIVARP47762_P050
Anti-MAFA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAFA (v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR