Anti-EIF3M Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP48293_T100
AVIVARP48293_T100EA 349 EUR
AVIVARP48293_T100
Anti-EIF3M Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EIF3M (eukaryotic translation initiation factor 3, subunit M)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen