Anti-KLHDC8B Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP48449_P050
AVIVARP48449_P050EA 401 EUR
AVIVARP48449_P050
Anti-KLHDC8B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: KLHDC8B (kelch domain containing 8B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen