Anti-NSUN3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP48884_P050
AVIVARP48884_P050EA 401 EUR
AVIVARP48884_P050
Anti-NSUN3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NSUN3 (NOP2/Sun domain family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen