Anti-ECHDC3 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP48911_T100
AVIVARP48911_T100EA 349 EUR
AVIVARP48911_T100
Anti-ECHDC3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ECHDC3 (enoyl CoA hydratase domain containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR