Anti-QTRT1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP48946_P050
AVIVARP48946_P050EA 401 EUR
AVIVARP48946_P050
Anti-QTRT1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: QTRT1 (queuine tRNA-ribosyltransferase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen