Anti-GALNT16 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP49540_P050
AVIVARP49540_P050EA 401 EUR
AVIVARP49540_P050
Anti-GALNT16 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GALNT16 (UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 16)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen