Anti-XYLT2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP49702_P050
AVIVARP49702_P050EA 401 EUR
AVIVARP49702_P050
Anti-XYLT2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: XYLT2 (xylosyltransferase II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen