Anti-XYLT2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP49703_P050
AVIVARP49703_P050EA 401 EUR
AVIVARP49703_P050
Anti-XYLT2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: XYLT2 (xylosyltransferase II)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

TECHNISCHE DATEN

Antigen-Symbol XYLT2
Environmentally Preferable
Antigen-Name xylosyltransferase II
Antigen-Synonyme XT-II|XT2|SOS|xylT-II|PXYLT2
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Unconjugated
Reaktivität
Wirt Rabbit
Gen-ID 64132
Isotyp
Western blot Yes
Epitop C-terminal
Kreuzadsorption none
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR